Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYT1L SIRNA Assays ›
Gene Symbol:
MYT1L
Gene Name:
myelin transcription factor 1 like
Gene Aliases:
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location:
Chr.2: 1789113 - 2331388 on Build GRCh38
UniGene ID:
Hs.434418
Species:
Human
siRNA ID
HSS118079
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYT1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303052.1
NP_001289981.1
17
3412
NM_015025.3
NP_055840.2
17
3406
XM_011510318.1
XP_011508620.1
17
3474
XM_011510319.1
XP_011508621.1
17
3584
XM_011510320.2
XP_011508622.1
17
3515
XM_011510321.2
XP_011508623.1
17
3465
XM_011510322.2
XP_011508624.1
17
3278
XM_011510323.1
XP_011508625.1
16
3197
XM_011510324.1
XP_011508626.1
17
3474
XM_011510325.1
XP_011508627.1
17
3468
XM_011510326.1
XP_011508628.1
17
3474
XM_011510327.1
XP_011508629.1
17
3474
XM_011510328.2
XP_011508630.1
17
3474
XM_011510331.2
XP_011508633.1
17
3474
XM_011510332.2
XP_011508634.1
17
3474
XM_017003604.1
XP_016859093.1
18
3563
XM_017003605.1
XP_016859094.1
17
3142
XM_017003606.1
XP_016859095.1
17
3578
XM_017003607.1
XP_016859096.1
17
3509
XM_017003608.1
XP_016859097.1
17
3474
XM_017003609.1
XP_016859098.1
17
3459
XM_017003610.1
XP_016859099.1
18
3558
XM_017003611.1
XP_016859100.1
17
3468
XM_017003612.1
XP_016859101.1
17
3578
XM_017003613.1
XP_016859102.1
17
3578
XM_017003614.1
XP_016859103.1
17
3417
XM_017003615.1
XP_016859104.1
17
3474
XM_017003616.1
XP_016859105.1
17
3474
XM_017003617.1
XP_016859106.1
17
3468
XM_017003618.1
XP_016859107.1
17
3468
XM_017003619.1
XP_016859108.1
17
3578
XM_017003620.1
XP_016859109.1
10
2464
XM_017003621.1
XP_016859110.1
17
3474
XM_017003622.1
XP_016859111.1
17
3474
GenBank mRNA
AB029029.2
-
18
3191
AF036943.1
-
17
3307
AK299571.1
-
13
2629
AL133024.1
-
8
1467
BC031690.1
-
9
2155
BC137272.1
-
17
3209
9021269
BC137273.1
-
17
3209
9021270
BC150281.1
-
18
3191
9007267
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nervous system development
cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0