Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYCBP2 SIRNA Assays ›
Gene Symbol:
MYCBP2
Gene Name:
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases:
Myc-bp2, PAM, Phr
Chromosome Location:
Chr.13: 77044657 - 77327066 on Build GRCh38
UniGene ID:
Hs.591221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118141
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYCBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015057.4
NP_055872.4
13
2134
XM_005266299.2
XP_005266356.1
13
2158
XM_006719779.2
XP_006719842.1
13
2158
XM_006719780.2
XP_006719843.1
13
2158
XM_006719781.2
XP_006719844.1
13
2158
XM_006719782.2
XP_006719845.1
13
2158
XM_006719783.2
XP_006719846.1
13
2158
XM_006719784.2
XP_006719847.1
13
2158
XM_006719785.2
XP_006719848.1
13
2158
XM_006719786.2
XP_006719849.1
13
2158
XM_006719787.2
XP_006719850.1
13
2158
XM_006719788.2
XP_006719851.1
13
2158
XM_006719789.2
XP_006719852.1
13
2158
XM_011535004.2
XP_011533306.1
12
1697
XM_017020454.1
XP_016875943.1
13
2158
XM_017020455.1
XP_016875944.1
13
2158
XM_017020456.1
XP_016875945.1
13
2158
XM_017020457.1
XP_016875946.1
13
2158
XM_017020458.1
XP_016875947.1
13
2158
XM_017020459.1
XP_016875948.1
13
2158
XM_017020460.1
XP_016875949.1
13
2158
XM_017020461.1
XP_016875950.1
13
2158
XM_017020462.1
XP_016875951.1
13
2158
XM_017020463.1
XP_016875952.1
13
2158
XM_017020464.1
XP_016875953.1
13
2158
XM_017020465.1
XP_016875954.1
13
2158
XM_017020466.1
XP_016875955.1
13
2158
XM_017020467.1
XP_016875956.1
13
2158
XM_017020468.1
XP_016875957.1
12
1645
GenBank mRNA
AF075587.1
-
14
1899
BE886946.1
-
4
413
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0