Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYCBP2 SIRNA Assays ›
          Gene Symbol:
          MYCBP2
          Gene Name:
          MYC binding protein 2, E3 ubiquitin protein ligase
          Gene Aliases:
          Myc-bp2, PAM, Phr
          Chromosome Location:
          Chr.13: 77044657 - 77327066 on Build GRCh38
          UniGene ID:
          Hs.591221
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118143
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYCBP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015057.4 NP_055872.4 34, 35 5401
          XM_005266299.2 XP_005266356.1 34, 35 5425
          XM_006719779.2 XP_006719842.1 34, 35 5425
          XM_006719780.2 XP_006719843.1 34, 35 5425
          XM_006719781.2 XP_006719844.1 34, 35 5425
          XM_006719782.2 XP_006719845.1 34, 35 5425
          XM_006719783.2 XP_006719846.1 34, 35 5425
          XM_006719784.2 XP_006719847.1 33, 34 5317
          XM_006719785.2 XP_006719848.1 34, 35 5425
          XM_006719786.2 XP_006719849.1 34, 35 5425
          XM_006719787.2 XP_006719850.1 34, 35 5425
          XM_006719788.2 XP_006719851.1 34, 35 5425
          XM_006719789.2 XP_006719852.1 34, 35 5425
          XM_011535004.2 XP_011533306.1 33, 34 4964
          XM_011535005.2 XP_011533307.1 17, 18 2505
          XM_017020454.1 XP_016875943.1 34, 35 5425
          XM_017020455.1 XP_016875944.1 34, 35 5425
          XM_017020456.1 XP_016875945.1 34, 35 5425
          XM_017020457.1 XP_016875946.1 34, 35 5425
          XM_017020458.1 XP_016875947.1 33, 34 5317
          XM_017020459.1 XP_016875948.1 33, 34 5317
          XM_017020460.1 XP_016875949.1 34, 35 5425
          XM_017020461.1 XP_016875950.1 34, 35 5425
          XM_017020462.1 XP_016875951.1 34, 35 5425
          XM_017020463.1 XP_016875952.1 33, 34 5317
          XM_017020464.1 XP_016875953.1 34, 35 5425
          XM_017020465.1 XP_016875954.1 33, 34 5317
          XM_017020466.1 XP_016875955.1 34, 35 5425
          XM_017020467.1 XP_016875956.1 34, 35 5425
          XM_017020468.1 XP_016875957.1 33, 34 4912
          GenBank mRNA AF075587.1 - 35, 36 5166

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein ubiquitination
          protein binding
          zinc ion binding
          ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0