Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 35 | 6879 | ||
XM_011514422.1 | XP_011512724.1 | 34 | 7035 | |||
XM_011514423.2 | XP_011512725.1 | 35 | 7137 | |||
XM_011514424.1 | XP_011512726.1 | 34 | 6954 | |||
XM_011514425.1 | XP_011512727.1 | 32 | 6705 | |||
XM_011514426.1 | XP_011512728.1 | 34 | 7035 | |||
XM_011514427.1 | XP_011512729.1 | 30 | 5654 | |||
XM_011514429.1 | XP_011512731.1 | 27 | 4859 | |||
XM_017010589.1 | XP_016866078.1 | 34 | 7746 | |||
XM_017010590.1 | XP_016866079.1 | 28 | 5208 | |||
GenBank mRNA | AB014608.3 | - | 27 | 4513 | ||
AJ318215.1 | - | 34 | 6804 | |||
AK128364.1 | - | 1 | 232 | |||
AY145132.1 | - | 34 | 6804 | |||
BC002879.2 | - | 12 | 2145 | |||
CR749511.1 | - | 34 | 7258 | |||
CR749841.1 | - | 35 | 6794 |