Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPIN1 SIRNA Assays ›
Gene Symbol:
LPIN1
Gene Name:
lipin 1
Gene Aliases:
PAP1
Chromosome Location:
Chr.2: 11677544 - 11827409 on Build GRCh38
UniGene ID:
Hs.467740
Species:
Human
siRNA ID
HSS118307
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261427.1
NP_001248356.1
4
517
NM_001261428.1
NP_001248357.1
4
438
NM_001261429.1
NP_001248358.1
4
620
NM_145693.2
NP_663731.1
3
307
XM_006711869.2
XP_006711932.1
4
473
XM_006711870.3
XP_006711933.1
4
604
XM_006711871.2
XP_006711934.1
3
307
XM_006711872.2
XP_006711935.1
3
531
XM_006711874.2
XP_006711937.1
4
2135
XM_011510333.2
XP_011508635.1
4
473
XM_011510334.2
XP_011508636.1
4
500
XM_011510335.2
XP_011508637.1
6
963
XM_011510336.2
XP_011508638.1
5
845
XM_017003623.1
XP_016859112.1
4
337
XM_017003624.1
XP_016859113.1
4
532
XM_017003625.1
XP_016859114.1
3
249
XM_017003626.1
XP_016859115.1
3
307
XM_017003627.1
XP_016859116.1
6
963
XM_017003628.1
XP_016859117.1
6
963
XM_017003629.1
XP_016859118.1
4
532
XM_017003630.1
XP_016859119.1
3
249
XM_017003631.1
XP_016859120.1
2
231
XM_017003632.1
XP_016859121.1
2
231
GenBank mRNA
AK290235.1
-
3
267
AK294742.1
-
4
422
AK302922.1
-
4
496
BC030537.1
-
3
300
3906281
BP192775.1
-
4
423
D80010.1
-
3
250
DB153243.1
-
3
307
DC334711.1
-
4
439
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
triglyceride mobilization
phosphatidylethanolamine biosynthetic process
phosphatidylcholine biosynthetic process
mitotic nuclear envelope disassembly
fatty acid catabolic process
dephosphorylation
triglyceride biosynthetic process
organ regeneration
cellular response to insulin stimulus
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription coactivator activity
phosphatidate phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0