Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF9 SIRNA Assays ›
Gene Symbol:
ARHGEF9
Gene Name:
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases:
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
Chromosome Location:
Chr.X: 63634967 - 63786025 on Build GRCh38
UniGene ID:
Hs.54697
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118406
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173479.1
NP_001166950.1
5
1069
NM_001173480.1
NP_001166951.1
5
1475
NM_015185.2
NP_056000.1
6
1666
XM_005262249.2
XP_005262306.1
6
1728
XM_005262250.3
XP_005262307.1
6
1626
XM_005262251.1
XP_005262308.1
6
976
XM_005262252.1
XP_005262309.1
7
995
XM_011530890.1
XP_011529192.1
7
1410
XM_011530892.1
XP_011529194.1
6
1234
XM_017029363.1
XP_016884852.1
7
1755
XM_017029364.1
XP_016884853.1
8
1532
XM_017029365.1
XP_016884854.1
6
1633
XM_017029366.1
XP_016884855.1
7
1414
XM_017029367.1
XP_016884856.1
7
1403
XM_017029368.1
XP_016884857.1
7
1704
XM_017029369.1
XP_016884858.1
6
1728
XM_017029370.1
XP_016884859.1
6
1637
XM_017029371.1
XP_016884860.1
6
1626
XM_017029372.1
XP_016884861.1
6
976
XM_017029373.1
XP_016884862.1
7
1769
XM_017029374.1
XP_016884863.1
5
1457
XM_017029375.1
XP_016884864.1
6
1634
XM_017029376.1
XP_016884865.1
6
1634
XM_017029377.1
XP_016884866.1
6
1637
XM_017029378.1
XP_016884867.1
6
1728
XM_017029379.1
XP_016884868.1
6
976
XM_017029380.1
XP_016884869.1
5
1457
GenBank mRNA
AB007884.1
-
6
1628
AK289993.1
-
6
1080
AK294721.1
-
2
185
AK295033.1
-
5
1081
AK295178.1
-
6
976
AL832116.1
-
6
1237
BC117406.1
-
6
977
40125957
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_023957, AJ302676, AJ250425.
This siRNA also targets the following transcripts in Mouse: NM_001290385, NM_001290384, NM_001033329, AK122280, AK038840, and others.
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
ion transmembrane transport
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0