Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SCFD1 SIRNA Assays ›
          Gene Symbol:
          SCFD1
          Gene Name:
          sec1 family domain containing 1
          Gene Aliases:
          C14orf163, RA410, SLY1, SLY1P, STXBP1L2
          Chromosome Location:
          Chr.14: 30622254 - 30735828 on Build GRCh38
          UniGene ID:
          Hs.369168
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118462
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SCFD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257376.1 NP_001244305.1 2, 3 241
          NM_001283031.1 NP_001269960.1 4, 5 403
          NM_001283032.1 NP_001269961.1 3, 4 314
          NM_016106.3 NP_057190.2 4, 5 403
          NM_182835.2 NP_878255.1 3, 4 332
          XM_005267469.2 XP_005267526.1 4, 5 401
          XM_011536586.2 XP_011534888.1 4, 5 400
          XM_011536587.2 XP_011534889.1 4, 5 400
          XM_011536588.2 XP_011534890.1 4, 5 400
          XM_011536589.1 XP_011534891.1 4, 5 400
          XM_017021113.1 XP_016876602.1 4, 5 400
          GenBank mRNA AB020724.2 - 4, 5 338
          AF086916.1 - 4, 5 326
          AF092139.1 - 4, 5 319
          AF110646.1 - 4, 5 314
          AF319958.1 - 4, 5 342
          AK222820.1 - 4, 5 322
          AK290410.1 - 4, 5 338
          AK297873.1 - 3, 4 267
          AK298622.1 - 2, 3 228
          AK299222.1 - 4, 5 330
          AK301406.1 - 3, 4 249
          AK316212.1 - 3, 4 267
          BC017734.1 - 4, 5 318 4420306
          BG028643.1 - 3, 4 245
          BX325898.2 - 1, 2 94
          CA454055.1 - 4, 5 329
          DA151789.1 - 4, 5 401
          DB105324.1 - 3, 4 332

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          response to hypoxia
          retrograde vesicle-mediated transport, Golgi to ER
          post-Golgi vesicle-mediated transport
          vesicle docking involved in exocytosis
          phagocytosis
          response to toxic substance
          protein transport
          COPII vesicle coating
          regulation of protein transport
          regulation of ER to Golgi vesicle-mediated transport
          toxin transport
          negative regulation of autophagosome assembly
          protein binding
          syntaxin binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0