Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286715.1 | NP_001273644.1 | 13 | 1786 | ||
NM_001286717.1 | NP_001273646.1 | 13 | 1666 | |||
NM_015239.2 | NP_056054.2 | 14 | 1731 | |||
XM_005251847.3 | XP_005251904.1 | 14 | 1797 | |||
XM_005251848.2 | XP_005251905.1 | 14 | 1975 | |||
XM_005251849.3 | XP_005251906.1 | 13 | 1721 | |||
XM_011518418.2 | XP_011516720.1 | 14 | 1797 | |||
XM_011518420.2 | XP_011516722.1 | 4 | 5050 | |||
XM_011518421.2 | XP_011516723.1 | 14 | 1797 | |||
XM_017014544.1 | XP_016870033.1 | 14 | 2342 | |||
XM_017014545.1 | XP_016870034.1 | 14 | 1677 | |||
XM_017014546.1 | XP_016870035.1 | 13 | 1564 | |||
XM_017014547.1 | XP_016870036.1 | 13 | 1721 | |||
XM_017014548.1 | XP_016870037.1 | 13 | 1664 | |||
XM_017014549.1 | XP_016870038.1 | 12 | 1683 | |||
XM_017014550.1 | XP_016870039.1 | 14 | 1797 | |||
GenBank mRNA | AB028958.2 | - | 14 | 1709 | ||
AK001544.1 | - | 15 | 1531 | |||
AK022562.1 | - | 2 | 374 | |||
AK023280.1 | - | 2 | 366 | |||
AK295304.1 | - | 10 | 1225 | |||
AK295774.1 | - | 13 | 1666 | |||
AK299506.1 | - | 10 | 1226 | |||
AL833359.1 | - | 2 | 323 | |||
BC060815.1 | - | 14 | 1696 | 30344122 | ||
BX648366.1 | - | 2 | 778 |