Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015245.2 | NP_056060.2 | 22 | 3378 | ||
XM_005248964.3 | XP_005249021.1 | 23 | 3429 | |||
XM_006715036.2 | XP_006715099.1 | 23 | 3432 | |||
XM_011514431.2 | XP_011512733.1 | 23 | 3432 | |||
XM_011514432.2 | XP_011512734.1 | 23 | 3429 | |||
XM_011514433.1 | XP_011512735.1 | 23 | 3432 | |||
XM_011514434.2 | XP_011512736.1 | 22 | 3369 | |||
XM_011514435.2 | XP_011512737.1 | 22 | 3366 | |||
XM_011514436.2 | XP_011512738.1 | 23 | 7001 | |||
XM_011514437.2 | XP_011512739.1 | 13 | 3257 | |||
XM_017010593.1 | XP_016866082.1 | 13 | 1855 | |||
GenBank mRNA | BC031934.1 | - | 24 | 3044 | 4819948 | |
BC049213.1 | - | 11 | 1221 | |||
BC132832.1 | - | 22 | 3315 | 40146854 | ||
BX094969.1 | - | 5 | 507 | |||
D86982.1 | - | 22 | 3375 |