Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM1 SIRNA Assays ›
Gene Symbol:
R3HDM1
Gene Name:
R3H domain containing 1
Gene Aliases:
R3HDM
Chromosome Location:
Chr.2: 135531429 - 135725270 on Build GRCh38
UniGene ID:
Hs.412462
Species:
Human
siRNA ID
HSS118878
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282798.1
NP_001269727.1
3
448
NM_001282799.1
NP_001269728.1
3
443
NM_015361.3
NP_056176.2
3
443
XM_005263626.3
XP_005263683.1
3
432
XM_005263627.3
XP_005263684.1
3
432
XM_005263628.3
XP_005263685.1
3
432
XM_005263629.3
XP_005263686.1
3
432
XM_006712389.2
XP_006712452.1
3
432
XM_011510870.1
XP_011509172.1
3
432
XM_011510871.1
XP_011509173.1
3
448
XM_011510872.2
XP_011509174.1
3
432
XM_011510873.2
XP_011509175.1
3
432
XM_011510874.2
XP_011509176.1
3
432
XM_011510875.2
XP_011509177.1
3
448
XM_011510876.2
XP_011509178.1
3
432
XM_011510877.2
XP_011509179.1
3
432
XM_011510878.2
XP_011509180.1
3
432
XM_011510879.2
XP_011509181.1
3
432
XM_011510881.1
XP_011509183.1
3
432
XM_017003718.1
XP_016859207.1
3
448
XM_017003719.1
XP_016859208.1
3
469
XM_017003720.1
XP_016859209.1
3
448
XM_017003721.1
XP_016859210.1
3
448
XM_017003722.1
XP_016859211.1
3
432
XM_017003723.1
XP_016859212.1
3
448
XM_017003724.1
XP_016859213.1
3
432
XM_017003725.1
XP_016859214.1
3
431
XM_017003726.1
XP_016859215.1
3
432
XM_017003727.1
XP_016859216.1
3
432
XM_017003728.1
XP_016859217.1
3
432
XM_017003729.1
XP_016859218.1
3
448
XM_017003730.1
XP_016859219.1
3
324
XM_017003731.1
XP_016859220.1
3
430
XM_017003732.1
XP_016859221.1
3
430
XM_017003733.1
XP_016859222.1
3
434
XM_017003734.1
XP_016859223.1
3
432
XM_017003735.1
XP_016859224.1
3
324
XM_017003736.1
XP_016859225.1
3
429
GenBank mRNA
AK290015.1
-
3
334
BC001217.1
-
3
425
BC041093.1
-
3
385
5556247
D21852.1
-
3
397
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0