Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167614.1 | NP_001161086.1 | 9 | 2186 | ||
NM_014319.4 | NP_055134.2 | 9 | 2189 | |||
GenBank mRNA | AF112299.2 | - | 9 | 2169 | ||
AK024954.1 | - | 9 | 864 | |||
AK295406.1 | - | 3 | 237 | |||
AK312473.1 | - | 9 | 2186 | |||
AL137533.1 | - | 1 | 25 |