Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012198.3 | NP_036330.1 | 5, 6 | 639 | ||
XM_005246446.2 | XP_005246503.1 | 5, 6 | 580 | |||
XM_006712398.3 | XP_006712461.1 | 5, 6 | 812 | |||
XM_006712400.3 | XP_006712463.1 | 6, 7 | 890 | |||
XM_006712401.3 | XP_006712464.1 | 5, 6 | 891 | |||
XM_011510926.2 | XP_011509228.1 | 5, 6 | 591 | |||
XM_011510927.2 | XP_011509229.1 | 5, 6 | 586 | |||
XM_011510928.2 | XP_011509230.1 | 3, 4 | 524 | |||
XM_017003764.1 | XP_016859253.1 | 6, 7 | 903 | |||
XM_017003765.1 | XP_016859254.1 | 6, 7 | 989 | |||
XM_017003766.1 | XP_016859255.1 | 6, 7 | 786 | |||
XM_017003767.1 | XP_016859256.1 | 5, 6 | 662 | |||
XM_017003768.1 | XP_016859257.1 | 5, 6 | 657 | |||
GenBank mRNA | AB209768.1 | - | 5, 6 | 528 | ||
AK295413.1 | - | 5, 6 | 572 | |||
AK312349.1 | - | 5, 6 | 588 | |||
BC005214.1 | - | 5, 6 | 562 | 3996889 | ||
M81637.1 | - | 5, 6 | 582 |