Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABI3BP SIRNA Assays ›
          Gene Symbol:
          ABI3BP
          Gene Name:
          ABI family member 3 binding protein
          Gene Aliases:
          NESHBP, TARSH
          Chromosome Location:
          Chr.3: 100749335 - 100993508 on Build GRCh38
          UniGene ID:
          Hs.477015
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS119461
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABI3BP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015429.3 NP_056244.2 12 1193
          XM_005247281.2 XP_005247338.1 14 1343
          XM_005247282.2 XP_005247339.1 14 1343
          XM_005247283.2 XP_005247340.1 14 1343
          XM_005247284.2 XP_005247341.1 14 1343
          XM_005247285.2 XP_005247342.1 14 1343
          XM_005247287.2 XP_005247344.1 14 1343
          XM_005247288.2 XP_005247345.1 13 1271
          XM_005247289.2 XP_005247346.1 14 1343
          XM_005247290.2 XP_005247347.1 14 1343
          XM_005247291.2 XP_005247348.1 14 1343
          XM_005247293.2 XP_005247350.1 14 1343
          XM_005247294.2 XP_005247351.1 14 1343
          XM_005247295.2 XP_005247352.1 14 1343
          XM_005247296.2 XP_005247353.1 14 1343
          XM_005247298.2 XP_005247355.1 14 1343
          XM_005247299.3 XP_005247356.1 12 1211
          XM_005247300.2 XP_005247357.1 14 1343
          XM_005247301.2 XP_005247358.1 14 1343
          XM_005247302.2 XP_005247359.1 14 1343
          XM_005247303.2 XP_005247360.1 14 1343
          XM_005247305.2 XP_005247362.1 14 1343
          XM_005247306.2 XP_005247363.1 14 1343
          XM_005247309.1 XP_005247366.1 12 976
          XM_006713569.2 XP_006713632.1 14 1343
          XM_006713570.2 XP_006713633.1 14 1343
          XM_006713571.2 XP_006713634.1 14 1343
          XM_006713572.2 XP_006713635.1 11 1121
          XM_011512639.1 XP_011510941.1 14 1343
          XM_011512640.1 XP_011510942.1 14 1343
          XM_011512641.1 XP_011510943.1 13 1268
          XM_011512643.1 XP_011510945.1 14 1343
          XM_011512644.1 XP_011510946.1 14 1343
          XM_011512645.1 XP_011510947.1 14 1343
          XM_011512646.1 XP_011510948.1 14 1343
          XM_011512647.1 XP_011510949.1 14 1343
          XM_011512648.1 XP_011510950.1 13 1271
          XM_011512649.1 XP_011510951.1 14 1343
          XM_011512651.1 XP_011510953.1 14 1343
          XM_011512652.1 XP_011510954.1 12 1196
          XM_011512653.1 XP_011510955.1 14 1343
          XM_011512654.1 XP_011510956.1 14 1343
          XM_011512657.1 XP_011510959.1 14 1343
          XM_011512658.1 XP_011510960.1 14 1343
          XM_011512659.1 XP_011510961.1 14 1343
          XM_011512660.1 XP_011510962.1 12 1196
          XM_017006105.1 XP_016861594.1 14 1343
          XM_017006106.1 XP_016861595.1 14 1343
          XM_017006107.1 XP_016861596.1 14 1343
          XM_017006108.1 XP_016861597.1 14 1343
          GenBank mRNA AL833204.1 - 14 1303
          BC030221.1 - 12 1173 5205716

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          collagen binding
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0