Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIN3A SIRNA Assays ›
          Gene Symbol:
          SIN3A
          Gene Name:
          SIN3 transcription regulator family member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 75369379 - 75455819 on Build GRCh38
          UniGene ID:
          Hs.513039
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS119557
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SIN3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145357.1 NP_001138829.1 13 2167
          NM_001145358.1 NP_001138830.1 13 2342
          NM_015477.2 NP_056292.1 13 2244
          XM_006720465.3 XP_006720528.1 13 2050
          XM_006720466.3 XP_006720529.1 14 2440
          XM_006720467.3 XP_006720530.1 15 2258
          GenBank mRNA AF418569.1 - 13 2244
          AK027559.1 - 13 2244
          AK096477.1 - 13 2293
          AL832463.1 - 13 2168
          BC137098.1 - 13 2095 9021095
          BC137099.1 - 13 2095 9021096
          BX649162.1 - 13 2281

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          activation of innate immune response
          positive regulation of defense response to virus by host
          hematopoietic progenitor cell differentiation
          DNA replication
          transcription, DNA-templated
          protein deacetylation
          aging
          regulation of hormone levels
          positive regulation of G2/M transition of mitotic cell cycle
          histone deacetylation
          positive regulation of chromatin silencing
          cellular protein localization
          negative regulation of circadian rhythm
          negative regulation of apoptotic process
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          cellular lipid metabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          response to methylglyoxal
          cellular response to glucose stimulus
          negative regulation of protein localization to nucleus
          negative regulation of histone H3-K27 acetylation
          cellular response to dopamine
          negative regulation of transcription regulatory region DNA binding
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II repressing transcription factor binding
          RNA polymerase II transcription corepressor activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          histone deacetylase activity
          protein binding
          protein complex binding
          protein deacetylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0