Search
Search

KRT23| Gene Symbol: KRT23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282433.1 | NP_001269362.1 | 7 | 1278 | ||
| NM_015515.4 | NP_056330.3 | 8 | 1806 | |||
| XM_005257200.4 | XP_005257257.1 | 7 | 859 | |||
| XM_011524595.1 | XP_011522897.1 | 7 | 849 | |||
| GenBank mRNA | AF102848.1 | - | 7 | 1234 | ||
| AK002047.1 | - | 7 | 1218 | |||
| AK225866.1 | - | 7 | 1282 | |||
| AK289449.1 | - | 7 | 1301 | |||
| AK302109.1 | - | 7 | 1087 | |||
| AK314604.1 | - | 7 | 1018 | |||
| AL117538.1 | - | 6 | 1062 | |||
| BC028356.2 | - | 8 | 1586 | 4838347 | ||
| CV373638.1 | - | 4 | 494 | |||