Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GIGYF2 SIRNA Assays ›
          Gene Symbol:
          GIGYF2
          Gene Name:
          GRB10 interacting GYF protein 2
          Gene Aliases:
          GYF2, PARK11, PERQ2, PERQ3, TNRC15
          Chromosome Location:
          Chr.2: 232697305 - 232860577 on Build GRCh38
          UniGene ID:
          Hs.565319
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS119752
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GIGYF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001103146.1 NP_001096616.1 14, 15 1834
          NM_001103147.1 NP_001096617.1 16, 17 1967
          NM_001103148.1 NP_001096618.1 13, 14 1750
          NM_015575.3 NP_056390.2 16, 17 1974
          NR_103492.1 13, 14 1750
          GenBank mRNA AB014542.2 - 16, 17 1926
          AK001739.1 - 16, 17 1948
          AK299418.1 - 15, 16 1878
          AK302998.1 - 8, 9 1336
          AY176045.1 - 16, 17 1948
          BC014047.2 - 1, 2 158
          BC043402.1 - 14, 15 1769
          BC136251.1 - 13, 14 1705 9020238
          BC143352.1 - 13, 14 1705
          BC146775.1 - 16, 17 1926 9007224
          BX537885.1 - 14, 15 1769
          BX538172.1 - 13, 14 1718
          BX538321.1 - 14, 15 1790
          BX647505.1 - 13, 14 1581

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          feeding behavior
          adult locomotory behavior
          post-embryonic development
          negative regulation of translation
          spinal cord motor neuron differentiation
          mitotic G1 DNA damage checkpoint
          multicellular organism growth
          cellular protein metabolic process
          insulin-like growth factor receptor signaling pathway
          homeostasis of number of cells within a tissue
          musculoskeletal movement
          neuromuscular process controlling balance
          cell-cell adhesion
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0