Search
Search

GBGT1| Gene Symbol: GBGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282629.1 | NP_001269558.1 | 7 | 974 | ||
| NM_001282632.1 | NP_001269561.1 | 6 | 942 | |||
| NM_001288572.1 | NP_001275501.1 | 7 | 1030 | |||
| NM_001288573.1 | NP_001275502.1 | 7 | 1030 | |||
| NM_021996.5 | NP_068836.2 | 7 | 993 | |||
| GenBank mRNA | AK074639.1 | - | 7 | 941 | ||
| AK074944.1 | - | 5 | 1956 | |||
| AK291498.1 | - | 7 | 962 | |||
| AK303825.1 | - | 6 | 911 | |||
| AK307687.1 | - | 7 | 999 | |||
| AK314097.1 | - | 7 | 962 | |||
| AY358175.1 | - | 7 | 942 | |||
| BC032499.1 | - | 7 | 945 | 5223113 | ||