Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014365.2 | NP_055180.1 | 1 | 857 | ||
GenBank mRNA | AF133207.1 | - | 1 | 694 | ||
AF191017.1 | - | 1 | 857 | |||
AF217987.1 | - | 1 | 702 | |||
AF250138.1 | - | 1 | 342 | |||
AK312501.1 | - | 1 | 710 | |||
AL136936.1 | - | 1 | 687 | |||
BC002673.2 | - | 1 | 825 | 3607117 | ||
BT006876.1 | - | 1 | 334 | |||
CR533453.1 | - | 1 | 334 |