Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003792.2 | NP_001003792.1 | 5 | 836 | ||
NM_001003793.2 | NP_001003793.1 | 5 | 839 | |||
NM_001177711.1 | NP_001171182.1 | 5 | 839 | |||
NM_001177712.1 | NP_001171183.1 | 5 | 839 | |||
NM_014483.3 | NP_055298.2 | 5 | 839 | |||
XM_005265060.2 | XP_005265117.1 | 5 | 1288 | |||
XM_005265061.2 | XP_005265118.1 | 5 | 1291 | |||
XM_005265062.1 | XP_005265119.1 | 5 | 1291 | |||
XM_005265063.1 | XP_005265120.1 | 2 | 496 | |||
XM_005265065.4 | XP_005265122.1 | 2 | 189 | |||
XM_017006178.1 | XP_016861667.1 | 5 | 1288 | |||
XM_017006179.1 | XP_016861668.1 | 5 | 1288 | |||
XM_017006180.1 | XP_016861669.1 | 5 | 1291 | |||
XM_017006181.1 | XP_016861670.1 | 5 | 1288 | |||
XM_017006182.1 | XP_016861671.1 | 6 | 1367 | |||
GenBank mRNA | AF023259.1 | - | 5 | 616 | ||
AK292789.1 | - | 5 | 515 | |||
AY236871.1 | - | 5 | 699 | |||
AY236872.1 | - | 5 | 696 | |||
BC117315.1 | - | 5 | 536 | 40125866 | ||
BC143521.1 | - | 5 | 536 | 9052030 |