Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOSIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270960.1 | NP_001257889.1 | 8 | 810 | ||
NM_015953.4 | NP_057037.1 | 9 | 929 | |||
XM_005258964.4 | XP_005259021.1 | 8 | 846 | |||
XM_006723235.3 | XP_006723298.1 | 8 | 880 | |||
XM_011527015.2 | XP_011525317.1 | 8 | 854 | |||
XM_011527017.2 | XP_011525319.1 | 8 | 886 | |||
XM_017026851.1 | XP_016882340.1 | 8 | 904 | |||
XM_017026852.1 | XP_016882341.1 | 8 | 885 | |||
GenBank mRNA | AF132959.1 | - | 8 | 795 | ||
AK092444.1 | - | 6 | 2005 | |||
AK291535.1 | - | 8 | 794 | |||
BC009299.2 | - | 8 | 780 | 4125665 | ||
BC010077.2 | - | 9 | 903 | 4309619 | ||
BC011249.1 | - | 7 | 764 | 3452886 |