Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015993.2 | NP_057077.1 | 2, 3 | 442 | ||
GenBank mRNA | AF137386.1 | - | 2, 3 | 438 | ||
AK130584.1 | - | 2, 3 | 434 | |||
AK289599.1 | - | 2, 3 | 419 | |||
AK313910.1 | - | 2, 3 | 442 | |||
BC002760.2 | - | 2, 3 | 405 | 3633345 | ||
DA366260.1 | - | 2, 3 | 443 |