Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303458.2 | NP_001290387.1 | 9, 10 | 1192 | ||
NM_001303459.2 | NP_001290388.1 | 9, 10 | 1192 | |||
NM_001323578.1 | NP_001310507.1 | 9, 10 | 1360 | |||
NM_001323579.1 | NP_001310508.1 | 10, 11 | 1330 | |||
NM_001323580.1 | NP_001310509.1 | 9, 10 | 1335 | |||
NM_001323581.1 | NP_001310510.1 | 10, 11 | 1305 | |||
NM_016004.4 | NP_057088.2 | 9, 10 | 1217 | |||
XM_017027863.1 | XP_016883352.1 | 10, 11 | 1104 | |||
GenBank mRNA | AF151811.1 | - | 9, 10 | 817 | ||
AK001436.1 | - | 9, 10 | 817 | |||
BC039831.1 | - | 9, 10 | 898 | 6095634 |