Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KMT5B SIRNA Assays ›
Gene Symbol:
KMT5B
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
CGI-85, CGI85, SUV420H1
Chromosome Location:
Chr.11: 68154863 - 68213904 on Build GRCh38
UniGene ID:
Hs.632120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS121687
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KMT5B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300907.1
NP_001287836.1
11
1383
NM_001300908.1
NP_001287837.1
10
1350
NM_001300909.1
NP_001287838.1
9
1334
NM_016028.4
NP_057112.3
10
1287
NM_017635.4
NP_060105.3
10
1315
XM_005274035.3
XP_005274092.2
10
1535
XM_005274036.3
XP_005274093.2
9
1466
XM_006718581.1
XP_006718644.1
9
1197
XM_011545091.1
XP_011543393.1
9
1109
XM_011545092.2
XP_011543394.1
9
1299
XM_011545093.2
XP_011543395.1
10
1580
XM_011545094.2
XP_011543396.1
10
1535
XM_017017876.1
XP_016873365.1
11
1896
XM_017017877.1
XP_016873366.1
11
1682
XM_017017878.1
XP_016873367.1
11
1627
XM_017017879.1
XP_016873368.1
11
1428
XM_017017880.1
XP_016873369.1
10
1535
GenBank mRNA
AF151843.1
-
9
1029
AK025183.1
-
9
975
AK025461.1
-
5
439
AK026428.1
-
5
424
AK299201.1
-
9
1163
AK314336.1
-
10
1177
BC002522.2
-
10
1234
BC012933.1
-
2
70
BC065287.1
-
10
1239
BC087834.1
-
10
1256
BC098121.1
-
9
1081
40001705
BC099714.1
-
9
1036
40001707
BC103498.1
-
9
1036
40001706
BC104483.1
-
9
1036
40001704
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108512, XM_008760150, XM_008760151.
This siRNA also targets the following transcripts in Mouse: NM_001167887, NM_001167889, NM_001167886, NM_144871, NM_001167885, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
muscle organ development
histone H4-K20 trimethylation
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0