Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CRBN SIRNA Assays ›
          Gene Symbol:
          CRBN TRNT1
          Gene Name:
          cereblon
          tRNA nucleotidyl transferase 1
          Gene Aliases:
          MRT2, MRT2A
          CCA1, CGI-47, MtCCA, RPEM, SIFD
          Chromosome Location:
          Chr.3: 3148490 - 3179717 on Build GRCh38
          Chromosome Location:
          Chr.3: 3126916 - 3153435 on Build GRCh38
          UniGene ID:
          Hs.18925
          Hs.732725
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS121807
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CRBN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001173482.1 NP_001166953.1 10 1093
          NM_016302.3 NP_057386.2 10 1096
          XM_005265202.3 XP_005265259.1 9 977
          XM_011533793.2 XP_011532095.1 11 1224
          XM_011533794.2 XP_011532096.1 10 1095
          GenBank mRNA AF117230.1 - 10 1059
          AK027507.1 - 10 1074
          AK058191.1 - 10 1070
          AK123077.1 - 2 1469
          AK301423.1 - 9 968
          BC017419.1 - 10 1084 4672890
          BC067811.1 - 11 1077 5264951
          BX647351.1 - 2 3408
          CR627060.1 - 6 3489
          Gene Symbol: TRNT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011533778.2 XP_011532080.1 9 3435

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease ubiquitin-protein ligase protein modifying enzyme RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          negative regulation of protein homooligomerization
          negative regulation of ion transmembrane transport
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of protein homodimerization activity
          tRNA 3'-terminal CCA addition
          protein targeting to mitochondrion
          tRNA 3'-end processing
          mitochondrial tRNA 3'-end processing
          ATP-dependent peptidase activity
          protein binding
          metal ion binding
          tRNA binding
          tRNA adenylyltransferase activity
          ATP binding
          RNA polymerase activity
          CTP:tRNA cytidylyltransferase activity
          CTP:3'-cytidine-tRNA cytidylyltransferase activity
          ATP:3'-cytidine-cytidine-tRNA adenylyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4tghz:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0