Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@6456c9d9
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
UCHL5
Gene Name:
ubiquitin C-terminal hydrolase L5
Gene Aliases:
CGI-70, INO80R, UCH-L5, UCH37
Chromosome Location:
Chr.1: 193012366 - 193060140 on Build GRCh38
UniGene ID:
Hs.145469
Species:
Human
siRNA ID
HSS122113
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price (USD)
401.00
Your Price (USD)
Online offer (USD):
401.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCHL5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199261.1
NP_001186190.1
1, 2
198
NM_001199262.1
NP_001186191.1
1, 2
198
NM_001199263.1
NP_001186192.1
1, 2
198
NM_015984.3
NP_057068.1
1, 2
198
XM_005245244.4
XP_005245301.1
1, 2
281
XM_005245246.4
XP_005245303.1
1, 2
281
XM_006711366.3
XP_006711429.1
1, 2
281
XM_006711367.3
XP_006711430.1
1, 2
281
XM_006711368.3
XP_006711431.1
1, 2
281
XM_006711369.2
XP_006711432.1
1, 2
281
XM_011509604.1
XP_011507906.1
1, 2
281
XM_011509608.2
XP_011507910.1
1, 2
172
XM_017001429.1
XP_016856918.1
1, 2
281
XM_017001430.1
XP_016856919.1
1, 2
281
XM_017001431.1
XP_016856920.1
1, 2
281
XM_017001432.1
XP_016856921.1
1, 2
281
XM_017001433.1
XP_016856922.1
1, 2
281
XM_017001436.1
XP_016856925.1
1, 2
172
XM_017001437.1
XP_016856926.1
1, 2
172
GenBank mRNA
AF147717.1
-
1, 2
196
AF151828.1
-
1, 2
78
AF157320.1
-
1, 2
213
AK024751.1
-
1, 2
144
AK225794.1
-
1, 2
131
AK296487.1
-
1, 2
171
AK314445.1
-
1, 2
142
BC015521.1
-
1, 2
131
3912584
BC025369.2
-
1, 2
176
3937407
BT006790.1
-
1, 2
65
DA255130.1
-
1, 2
198
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
hydrolase
protease
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-green:8080/10.253.229.154:8080.
git-commit: 0914912cb48844dc9118fffd0bcf8fc2c0fe1c71
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.17.0-2021.02.06-1.0