Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PEX5L SIRNA Assays ›
          Gene Symbol:
          PEX5L
          Gene Name:
          peroxisomal biogenesis factor 5 like
          Gene Aliases:
          PEX5R, PEX5RP, PXR2, PXR2B, TRIP8b
          Chromosome Location:
          Chr.3: 179794958 - 180037053 on Build GRCh38
          UniGene ID:
          Hs.235331
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS122344
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PEX5L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256750.1 NP_001243679.1 13, 14 1969
          NM_001256751.1 NP_001243680.1 12, 13 1903
          NM_001256752.1 NP_001243681.1 12, 13 1870
          NM_001256753.1 NP_001243682.1 11, 12 1798
          NM_001256754.1 NP_001243683.1 13, 14 1705
          NM_001256755.1 NP_001243684.1 12, 13 1510
          NM_001256756.1 NP_001243685.1 12, 13 1600
          NM_016559.2 NP_057643.1 13, 14 1975
          XM_011512882.2 XP_011511184.1 14, 15 2587
          XM_011512884.2 XP_011511186.1 14, 15 2583
          XM_011512885.2 XP_011511187.1 14, 15 1998
          XM_011512886.2 XP_011511188.1 13, 14 1804
          XM_011512887.2 XP_011511189.1 12, 13 1366
          XM_011512888.2 XP_011511190.1 12, 13 1666
          XM_011512891.2 XP_011511193.1 11, 12 2163
          XM_011512892.2 XP_011511194.1 10, 11 1969
          XM_017006601.1 XP_016862090.1 13, 14 2395
          XM_017006602.1 XP_016862091.1 13, 14 1800
          XM_017006603.1 XP_016862092.1 13, 14 2391
          XM_017006604.1 XP_016862093.1 11, 12 2981
          XM_017006605.1 XP_016862094.1 11, 12 1666
          XM_017006606.1 XP_016862095.1 13, 14 2472
          XM_017006607.1 XP_016862096.1 12, 13 3169
          XM_017006608.1 XP_016862097.1 12, 13 1819
          XM_017006609.1 XP_016862098.1 12, 13 2312
          XM_017006610.1 XP_016862099.1 11, 12 1747
          XM_017006611.1 XP_016862100.1 11, 12 2124
          GenBank mRNA AB032592.1 - 13, 14 1686
          AB032593.1 - 13, 14 1651
          AJ245503.1 - 6, 7 734
          AK294227.1 - 10, 11 1200
          AK294501.1 - 11, 12 1673
          AK295349.1 - 12, 13 1368
          AK295386.1 - 12, 13 1600
          AK299633.1 - 12, 13 1776
          AK303716.1 - 13, 14 1696
          AK314283.1 - 13, 14 1853
          BC036183.1 - 12, 13 1754 4793921

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into peroxisome matrix, docking
          regulation of cAMP-mediated signaling
          peroxisome targeting sequence binding
          small GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0