Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5L SIRNA Assays ›
Gene Symbol:
PEX5L
Gene Name:
peroxisomal biogenesis factor 5 like
Gene Aliases:
PEX5R, PEX5RP, PXR2, PXR2B, TRIP8b
Chromosome Location:
Chr.3: 179794958 - 180037053 on Build GRCh38
UniGene ID:
Hs.235331
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS122345
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256750.1
NP_001243679.1
15
2155
NM_001256751.1
NP_001243680.1
14
2089
NM_001256752.1
NP_001243681.1
14
2056
NM_001256753.1
NP_001243682.1
13
1984
NM_001256754.1
NP_001243683.1
15
1891
NM_001256755.1
NP_001243684.1
14
1696
NM_001256756.1
NP_001243685.1
14
1786
NM_016559.2
NP_057643.1
15
2161
XM_011512882.2
XP_011511184.1
16
2773
XM_011512884.2
XP_011511186.1
16
2769
XM_011512885.2
XP_011511187.1
16
2184
XM_011512886.2
XP_011511188.1
15
1990
XM_011512887.2
XP_011511189.1
14
1552
XM_011512888.2
XP_011511190.1
14
1852
XM_011512891.2
XP_011511193.1
13
2349
XM_011512892.2
XP_011511194.1
12
2155
XM_017006601.1
XP_016862090.1
15
2581
XM_017006602.1
XP_016862091.1
15
1986
XM_017006603.1
XP_016862092.1
15
2577
XM_017006604.1
XP_016862093.1
13
3167
XM_017006605.1
XP_016862094.1
13
1852
XM_017006606.1
XP_016862095.1
15
2658
XM_017006607.1
XP_016862096.1
14
3355
XM_017006608.1
XP_016862097.1
14
2005
XM_017006609.1
XP_016862098.1
14
2498
XM_017006610.1
XP_016862099.1
13
1933
XM_017006611.1
XP_016862100.1
13
2310
GenBank mRNA
AB032592.1
-
15
1872
AB032593.1
-
15
1837
AJ245503.1
-
8
920
AK294227.1
-
12
1386
AK294501.1
-
13
1859
AK295349.1
-
14
1554
AK295386.1
-
14
1786
AK299633.1
-
14
1962
AK303716.1
-
15
1882
AK314283.1
-
15
2039
BC036183.1
-
14
1940
4793921
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix, docking
regulation of cAMP-mediated signaling
peroxisome targeting sequence binding
small GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0