Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
siRNA ID
HSS122371
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
8
890
NM_016623.4
NP_057707.3
7
803
NR_046359.1
10
1156
NR_046360.1
10
1066
NR_046361.1
9
1056
XM_005250966.3
XP_005251023.1
6
659
XM_011517108.2
XP_011515410.1
9
952
XM_011517109.2
XP_011515411.1
9
724
XM_011517110.2
XP_011515412.1
8
864
XM_011517111.2
XP_011515413.1
8
682
XM_011517112.2
XP_011515414.1
10
907
XM_011517113.2
XP_011515415.1
9
918
XM_011517114.2
XP_011515416.1
9
824
XM_011517115.2
XP_011515417.1
8
831
XM_011517116.2
XP_011515418.1
9
865
XM_011517117.2
XP_011515419.1
8
834
XM_011517118.2
XP_011515420.1
8
778
XM_011517120.2
XP_011515422.1
7
777
XM_011517121.2
XP_011515423.1
10
789
XM_011517122.2
XP_011515424.1
8
868
XM_011517123.2
XP_011515425.1
8
641
XM_011517124.2
XP_011515426.1
7
756
XM_011517125.2
XP_011515427.1
7
695
XM_017013539.1
XP_016869028.1
10
911
XM_017013540.1
XP_016869029.1
10
770
XM_017013541.1
XP_016869030.1
8
898
XM_017013542.1
XP_016869031.1
10
948
XM_017013543.1
XP_016869032.1
10
1048
XM_017013544.1
XP_016869033.1
10
830
XM_017013545.1
XP_016869034.1
11
1094
XM_017013546.1
XP_016869035.1
9
861
XM_017013547.1
XP_016869036.1
11
953
XM_017013548.1
XP_016869037.1
9
957
XM_017013549.1
XP_016869038.1
9
820
XM_017013550.1
XP_016869039.1
8
871
XM_017013551.1
XP_016869040.1
10
866
XM_017013552.1
XP_016869041.1
10
911
XM_017013553.1
XP_016869042.1
9
724
XM_017013554.1
XP_016869043.1
11
936
XM_017013555.1
XP_016869044.1
8
891
XM_017013556.1
XP_016869045.1
8
637
XM_017013557.1
XP_016869046.1
9
683
XM_017013558.1
XP_016869047.1
9
828
XM_017013559.1
XP_016869048.1
9
687
XM_017013560.1
XP_016869049.1
7
802
XM_017013561.1
XP_016869050.1
7
554
XM_017013562.1
XP_016869051.1
8
741
GenBank mRNA
AF208851.1
-
9
742
AK001226.1
-
3
309
AK002059.1
-
10
866
AK130372.1
-
7
690
AY364259.1
-
4
357
AY598320.1
-
5
489
BC003599.1
-
8
669
3501828
BC016345.1
-
9
810
4041187
BC017297.2
-
10
1038
5017493
CR749628.1
-
8
636
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0