Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 2 | 361 | ||
GenBank mRNA | AF113537.1 | - | 2 | 230 | ||
AF123767.1 | - | 2 | 230 | |||
AF133422.1 | - | 2 | 214 | |||
AK294383.1 | - | 2 | 241 | |||
AK297196.1 | - | 2 | 245 | |||
AK312359.1 | - | 2 | 248 | |||
BC002619.2 | - | 2 | 244 | 3160345 | ||
BC018873.2 | - | 2 | 237 | |||
CR457142.1 | - | 1 | 99 | |||
DC366641.1 | - | 2 | 361 |