Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016024.2 | NP_057108.2 | 6 | 1052 | ||
GenBank mRNA | AF151837.1 | - | 5 | 987 | ||
AK292855.1 | - | 6 | 1034 | |||
AL050405.1 | - | 6 | 1027 | |||
BC033750.1 | - | 6 | 1020 | 5210341 | ||
BC040433.1 | - | 6 | 1003 |