Search
Search

HSPB11| Gene Symbol: HSPB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316935.1 | NP_001303864.1 | 5, 6 | 416 | ||
| NM_016126.3 | NP_057210.2 | 5, 6 | 613 | |||
| NR_133632.1 | 3, 4 | 437 | ||||
| NR_133633.1 | 6, 7 | 711 | ||||
| GenBank mRNA | AF100747.1 | - | 5, 6 | 385 | ||
| AF416725.1 | - | 4, 5 | 340 | |||
| AF417114.1 | - | 4, 5 | 340 | |||
| AK225916.1 | - | 5 | 2500 | |||
| AW024033.1 | - | 2, 3 | 187 | |||
| BC005245.1 | - | 5, 6 | 405 | 3838843 | ||
| BC045699.1 | - | 1 | 2202 | |||
| HY179788.1 | - | 5, 6 | 417 | |||
| L32869.1 | - | 3, 4 | 188 | |||
| X79302.1 | - | 3, 4 | 188 | |||