Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278666.1 | NP_001265595.1 | 7 | 819 | ||
NM_001278667.1 | NP_001265596.1 | 7 | 991 | |||
NM_001278668.1 | NP_001265597.1 | 7 | 818 | |||
NM_016277.4 | NP_057361.3 | 7 | 1288 | |||
NM_183227.2 | NP_899050.1 | 7 | 842 | |||
NR_103822.1 | 4 | 493 | ||||
GenBank mRNA | AB025427.1 | - | 6 | 634 | ||
AB034244.1 | - | 7 | 787 | |||
AF161486.1 | - | 7 | 784 | |||
AF498951.1 | - | 6 | 634 | |||
AK311123.1 | - | 4 | 482 | |||
AK313796.1 | - | 7 | 839 | |||
AY585189.1 | - | 6 | 634 | |||
BC015021.1 | - | 7 | 1254 | 3910708 | ||
CR749371.1 | - | 8 | 1022 |