Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERAP1 SIRNA Assays ›
          Gene Symbol:
          ERAP1
          Gene Name:
          endoplasmic reticulum aminopeptidase 1
          Gene Aliases:
          A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
          Chromosome Location:
          Chr.5: 96759245 - 96935983 on Build GRCh38
          UniGene ID:
          Hs.666524
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS122607
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040458.1 NP_001035548.1 4, 5 1143
          NM_001198541.1 NP_001185470.1 4, 5 901
          NM_016442.3 NP_057526.3 4, 5 1143
          XM_005272015.4 XP_005272072.1 4, 5 1094
          XM_005272016.4 XP_005272073.1 4, 5 1054
          XM_011543480.2 XP_011541782.1 8, 9 3363
          XM_011543481.2 XP_011541783.1 8, 9 3360
          XM_011543484.2 XP_011541786.1 8, 9 3359
          XM_011543485.2 XP_011541787.1 7, 8 3179
          XM_011543486.2 XP_011541788.1 8, 9 3363
          XM_017009580.1 XP_016865069.1 9, 10 3419
          XM_017009581.1 XP_016865070.1 7, 8 3205
          XM_017009582.1 XP_016865071.1 4, 5 878
          XM_017009583.1 XP_016865072.1 4, 5 1089
          GenBank mRNA AB011097.2 - 3, 4 817
          AF106037.1 - 4, 5 858
          AF183569.1 - 4, 5 863
          AF222340.1 - 4, 5 883
          AK291636.1 - 4, 5 893
          AK310535.1 - 3, 4 318
          AY028806.1 - 4, 5 893
          AY028807.1 - 4, 5 893
          AY358590.1 - 4, 5 881
          BC030775.1 - 4, 5 895 4824919
          DC398730.1 - 3, 4 361
          JX962699.1 - 3, 4 796
          JX962700.1 - 3, 4 796
          JX962701.1 - 3, 4 796
          JX962702.1 - 3, 4 796
          KM357870.1 - 3, 4 796
          KM357871.1 - 3, 4 796
          KM357872.1 - 3, 4 796
          KM357873.1 - 3, 4 796
          KM357874.1 - 3, 4 796
          KM357875.1 - 3, 4 796
          KM357876.1 - 3, 4 796
          KM357877.1 - 3, 4 796
          KM357878.1 - 3, 4 796
          KM357879.1 - 3, 4 796
          KM357880.1 - 3, 4 796
          KM357881.1 - 3, 4 796
          KM357882.1 - 3, 4 796
          KM357883.1 - 3, 4 796
          KM357884.1 - 3, 4 796
          KM357885.1 - 3, 4 796
          KM357886.1 - 3, 4 796
          KM357887.1 - 3, 4 796
          KM357888.1 - 3, 4 796
          KM357889.1 - 3, 4 796
          KM357890.1 - 3, 4 796
          KM357891.1 - 3, 4 796

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          adaptive immune response
          antigen processing and presentation of peptide antigen via MHC class I
          membrane protein ectodomain proteolysis
          regulation of blood pressure
          response to bacterium
          antigen processing and presentation of endogenous peptide antigen via MHC class I
          peptide catabolic process
          regulation of innate immune response
          fat cell differentiation
          positive regulation of angiogenesis
          endopeptidase activity
          aminopeptidase activity
          interleukin-6 receptor binding
          interleukin-1, Type II receptor binding
          tumor necrosis factor receptor binding
          protein binding
          metalloexopeptidase activity
          zinc ion binding
          peptide binding
          metalloaminopeptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-t6srt:80/100.66.79.227:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0