Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016578.3 | NP_057662.3 | 15 | 3807 | ||
XM_005274051.2 | XP_005274108.1 | 15 | 3833 | |||
XM_017017923.1 | XP_016873412.1 | 15 | 3581 | |||
GenBank mRNA | AF227948.2 | - | 10 | 3992 | ||
AF380176.1 | - | 15 | 3678 |