Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016578.3 | NP_057662.3 | 5 | 712 | ||
XM_005274051.2 | XP_005274108.1 | 5 | 747 | |||
XM_017017923.1 | XP_016873412.1 | 5 | 486 | |||
GenBank mRNA | AF059317.1 | - | 3 | 263 | ||
AF380176.1 | - | 5 | 583 | |||
BC015360.1 | - | 5 | 712 |