Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243270.1 | NP_001230199.1 | 18 | 2692 | ||
NM_001243271.1 | NP_001230200.1 | 19 | 2831 | |||
NM_014361.3 | NP_055176.1 | 19 | 2831 | |||
NM_175566.2 | NP_780775.1 | 16 | 2175 | |||
XM_011542871.1 | XP_011541173.1 | 18 | 2609 | |||
XM_017017926.1 | XP_016873415.1 | 20 | 2999 | |||
GenBank mRNA | AB013802.2 | - | 17 | 2406 | ||
AB013803.2 | - | 16 | 2184 | |||
AK294692.1 | - | 5 | 575 | |||
BC039255.1 | - | 19 | 2666 | 5273769 | ||
BC130619.1 | - | 18 | 2478 | 40146650 | ||
BC144185.1 | - | 18 | 2478 | 9052705 | ||
BC144186.1 | - | 19 | 2617 | 9052706 |