Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf62-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024622.1 | 3, 4 | 433 | |||
NR_024623.1 | 3, 4 | 421 | ||||
GenBank mRNA | AK096601.1 | - | 3, 4 | 421 | ||
BC117397.1 | - | 3, 4 | 315 | |||
BC117399.1 | - | 3, 4 | 327 | |||
DA776905.1 | - | 3, 4 | 421 |