Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018946.3 | NP_061819.2 | 6 | 1181 | ||
XM_011518787.2 | XP_011517089.1 | 6 | 937 | |||
XM_011518788.2 | XP_011517090.1 | 4 | 688 | |||
XM_017014811.1 | XP_016870300.1 | 5 | 882 | |||
GenBank mRNA | AF257466.1 | - | 6 | 1049 | ||
AK001659.1 | - | 6 | 1119 | |||
AK091184.1 | - | 4 | 2975 | |||
AK316608.1 | - | 6 | 1141 | |||
BC000008.2 | - | 6 | 1127 | 3505324 | ||
BC019315.1 | - | 6 | 1109 | 2821389 | ||
BQ446501.1 | - | 1 | 73 | |||
FJ224349.1 | - | 6 | 1049 |
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252320.3 | XP_005252377.1 | 7 | 2127 | ||
GenBank mRNA | AK123739.1 | - | 1 | 888 |