This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
UGT1A3
UGT1A4
- Gene Name:
-
UDP glucuronosyltransferase family 1 member A3
-
UDP glucuronosyltransferase family 1 member A4
- Gene Aliases:
-
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
-
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
- Chromosome Location:
-
Chr.2: 233729127 - 233773299 on Build GRCh38
- Chromosome Location:
-
Chr.2: 233718792 - 233773299 on Build GRCh38
- UniGene ID:
-
Hs.554822
-
Hs.554822
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
-
-
siRNA Details
|
Gene Symbol:
UGT1A3 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_019093.2 |
NP_061966.1 |
1
|
614
|
|
| GenBank mRNA |
AY435138.1 |
- |
1
|
614
|
|
|
Gene Symbol:
UGT1A4 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| GenBank mRNA |
BC121036.1 |
- |
1, 2
|
614
|
40117585 |
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System