Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017523.3 | NP_059993.2 | 6 | 805 | ||
NM_199139.2 | NP_954590.1 | 5 | 748 | |||
NR_046396.1 | 5 | 719 | ||||
NR_046397.1 | 4 | 662 | ||||
NR_046398.1 | 4 | 552 | ||||
XM_005256706.4 | XP_005256763.1 | 6 | 1377 | |||
XM_005256708.2 | XP_005256765.1 | 7 | 1367 | |||
XM_011523945.2 | XP_011522247.1 | 7 | 1192 | |||
XM_011523946.2 | XP_011522248.1 | 7 | 1192 | |||
XM_011523947.2 | XP_011522249.1 | 6 | 713 | |||
XM_011523948.1 | XP_011522250.1 | 6 | 1106 | |||
XM_017024780.1 | XP_016880269.1 | 6 | 1006 | |||
GenBank mRNA | AK056908.1 | - | 6 | 1601 | ||
AK290276.1 | - | 8 | 875 | |||
AK292710.1 | - | 6 | 585 | |||
AK292848.1 | - | 5 | 528 | |||
BC032776.1 | - | 4 | 335 | |||
BC058017.1 | - | 5 | 515 | |||
BC073156.1 | - | 6 | 604 | 6068243 | ||
BM475564.1 | - | 6 | 576 | |||
BX648588.1 | - | 5 | 1136 | |||
BX649188.1 | - | 6 | 3052 | |||
EF028165.1 | - | 5 | 477 | |||
EF028166.1 | - | 4 | 420 | |||
X99699.1 | - | 6 | 563 |