Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KLHL24 SIRNA Assays ›
Gene Symbol:
KLHL24
Gene Name:
kelch like family member 24
Gene Aliases:
DRE1, KRIP6
Chromosome Location:
Chr.3: 183635117 - 183684519 on Build GRCh38
UniGene ID:
Hs.407709
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS123273
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KLHL24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017644.3
NP_060114.2
3
782
XM_005247552.2
XP_005247609.1
4
873
XM_005247553.1
XP_005247610.1
4
827
XM_005247554.2
XP_005247611.1
3
738
XM_005247555.1
XP_005247612.1
3
810
XM_005247556.1
XP_005247613.1
2
732
XM_011512938.2
XP_011511240.1
2
674
XM_017006650.1
XP_016862139.1
3
1288
XM_017006651.1
XP_016862140.1
4
873
XM_017006652.1
XP_016862141.1
4
827
XM_017006653.1
XP_016862142.1
3
749
XM_017006654.1
XP_016862143.1
3
739
XM_017006655.1
XP_016862144.1
3
810
XM_017006656.1
XP_016862145.1
2
732
XM_017006657.1
XP_016862146.1
3
764
XM_017006658.1
XP_016862147.1
2
673
XM_017006659.1
XP_016862148.1
4
2574
XM_017006660.1
XP_016862149.1
3
764
XM_017006661.1
XP_016862150.1
4
867
XM_017006662.1
XP_016862151.1
3
798
XM_017006663.1
XP_016862152.1
3
787
GenBank mRNA
AK000066.1
-
1
164
AK026326.1
-
3
782
AY422472.1
-
3
782
BC142993.1
-
3
836
8860448
BX648466.1
-
4
874
BX648812.1
-
4
874
DQ925701.1
-
1
487
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of kainate selective glutamate receptor activity
ubiquitin-protein transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0