Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYM SIRNA Assays ›
Gene Symbol:
DYM
Gene Name:
dymeclin
Gene Aliases:
DMC, SMC
Chromosome Location:
Chr.18: 49041474 - 49460709 on Build GRCh38
UniGene ID:
Hs.162996
Species:
Human
siRNA ID
HSS123284
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017653.3
NP_060123.3
14
1892
XM_006722488.2
XP_006722551.1
14
1887
XM_006722490.2
XP_006722553.1
14
1887
XM_011526036.1
XP_011524338.1
14
1887
XM_011526037.1
XP_011524339.1
14
1889
XM_011526038.1
XP_011524340.1
14
1884
XM_011526039.1
XP_011524341.1
14
1887
XM_011526040.1
XP_011524342.1
15
1888
XM_011526041.1
XP_011524343.1
14
1887
XM_011526042.1
XP_011524344.1
14
1887
XM_017025795.1
XP_016881284.1
14
1886
XM_017025796.1
XP_016881285.1
15
1898
XM_017025797.1
XP_016881286.1
14
1825
XM_017025798.1
XP_016881287.1
14
1884
XM_017025799.1
XP_016881288.1
14
1887
XM_017025800.1
XP_016881289.1
14
1884
XM_017025801.1
XP_016881290.1
14
1886
GenBank mRNA
AK000078.1
-
10
1205
AK074611.1
-
14
1887
AK091256.1
-
15
1861
AK225205.1
-
10
1205
AK291303.1
-
14
1857
AK296579.1
-
9
1279
AK315091.1
-
15
1833
AL390156.1
-
3
194
AY364250.1
-
13
1527
BC001252.1
-
11
1291
BC064394.1
-
14
1874
5752466
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
bone development
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0