Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017706.4 | NP_060176.2 | 5 | 760 | ||
XM_005268469.2 | XP_005268526.1 | 5 | 760 | |||
XM_017009600.1 | XP_016865089.1 | 6 | 547 | |||
GenBank mRNA | AK000202.1 | - | 1 | 126 | ||
AK025355.1 | - | 5 | 707 | |||
AK056861.1 | - | 4 | 771 | |||
AK122862.1 | - | 3 | 2195 | |||
AK293417.1 | - | 2 | 247 | |||
BC002482.2 | - | 4 | 466 | |||
BC041687.1 | - | 5 | 642 | |||
BC068485.1 | - | 5 | 706 | 30334628 | ||
BQ069026.1 | - | 5 | 731 |