Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017748.4 | NP_060218.1 | 10 | 1415 | ||
NR_073428.1 | 11 | 1532 | ||||
GenBank mRNA | AK000298.1 | - | 10 | 1365 | ||
AK225373.1 | - | 10 | 1385 | |||
AK296115.1 | - | 11 | 1497 | |||
BC008833.2 | - | 10 | 1401 | 4304656 | ||
CR749362.1 | - | 6 | 597 |