Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017880.1 | NP_060350.1 | 3 | 638 | ||
XM_005264389.2 | XP_005264446.1 | 3 | 831 | |||
XM_005264390.2 | XP_005264447.1 | 3 | 700 | |||
XM_005264391.2 | XP_005264448.1 | 3 | 782 | |||
XM_011532937.2 | XP_011531239.1 | 2 | 576 | |||
XM_017004398.1 | XP_016859887.1 | 3 | 1119 | |||
XM_017004399.1 | XP_016859888.1 | 3 | 827 | |||
XM_017004400.1 | XP_016859889.1 | 3 | 711 | |||
GenBank mRNA | AK000565.1 | - | 3 | 638 | ||
AL136834.1 | - | 2 | 416 | |||
BC002825.2 | - | 3 | 627 | 3635986 | ||
BC005079.1 | - | 3 | 627 | 3615724 |