Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OXR1 SIRNA Assays ›
Gene Symbol:
OXR1
Gene Name:
oxidation resistance 1
Gene Aliases:
Nbla00307, TLDC3
Chromosome Location:
Chr.8: 106270178 - 106752694 on Build GRCh38
UniGene ID:
Hs.127286
Species:
Human
siRNA ID
HSS123840
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OXR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198532.1
NP_001185461.1
16
2635
NM_001198533.1
NP_001185462.1
17
2861
NM_001198534.1
NP_001185463.1
7
780
NM_001198535.1
NP_001185464.1
6
699
NM_018002.3
NP_060472.2
16
2780
NM_181354.4
NP_851999.2
14
2610
XM_006716595.2
XP_006716658.1
15
2549
XM_017013589.1
XP_016869078.1
15
2660
XM_017013590.1
XP_016869079.1
15
2501
XM_017013591.1
XP_016869080.1
15
2591
XM_017013592.1
XP_016869081.1
15
2338
XM_017013593.1
XP_016869082.1
15
2565
XM_017013594.1
XP_016869083.1
14
2484
XM_017013595.1
XP_016869084.1
14
2475
XM_017013596.1
XP_016869085.1
16
2648
XM_017013597.1
XP_016869086.1
14
2309
XM_017013598.1
XP_016869087.1
14
2380
XM_017013599.1
XP_016869088.1
13
2394
XM_017013600.1
XP_016869089.1
13
2251
GenBank mRNA
AB075503.1
-
7
726
AF309387.1
-
9
1078
AK000987.1
-
7
751
AK098168.1
-
1
1125
AK123667.1
-
6
1014
AK292899.1
-
14
2562
AK296561.1
-
16
2635
AK302844.1
-
7
728
AK303695.1
-
6
794
AK316153.1
-
6
633
AL833193.1
-
13
2191
BC032710.2
-
14
2552
5496492
BC035484.1
-
14
2564
FN650108.1
-
16
2671
FN650109.1
-
6
562
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to oxidative stress
adult walking behavior
negative regulation of neuron apoptotic process
oxidation-reduction process
cellular response to hydroperoxide
molecular_function
oxidoreductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0