Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018004.2 | NP_060474.1 | 5, 6 | 1054 | ||
XM_005247569.2 | XP_005247626.1 | 6, 7 | 1015 | |||
XM_017006715.1 | XP_016862204.1 | 6, 7 | 1175 | |||
GenBank mRNA | AB013911.1 | - | 4, 5 | 741 | ||
AK000996.1 | - | 5, 6 | 1054 | |||
AK129789.1 | - | 5, 6 | 904 | |||
AY450392.1 | - | 5, 6 | 1054 | |||
BC040355.1 | - | 6, 7 | 988 | 4748104 | ||
BG205309.1 | - | 1, 2 | 156 |