Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177663.1 | NP_001171134.1 | 22 | 2687 | ||
NM_001177664.1 | NP_001171135.1 | 22 | 2590 | |||
NM_001177665.1 | NP_001171136.1 | 21 | 2860 | |||
NM_018151.4 | NP_060621.3 | 22 | 2687 | |||
XM_005246665.3 | XP_005246722.1 | 22 | 2567 | |||
XM_011511395.2 | XP_011509697.1 | 21 | 2793 | |||
XM_017004422.1 | XP_016859911.1 | 22 | 2636 | |||
XM_017004423.1 | XP_016859912.1 | 21 | 2474 | |||
XM_017004424.1 | XP_016859913.1 | 21 | 2504 | |||
XM_017004425.1 | XP_016859914.1 | 20 | 2812 | |||
GenBank mRNA | AK001461.1 | - | 5 | 636 | ||
AK022932.1 | - | 17 | 2050 | |||
AK024033.1 | - | 6 | 793 | |||
AK299291.1 | - | 22 | 2564 | |||
AY584066.1 | - | 21 | 2526 | |||
AY585745.1 | - | 22 | 2636 | |||
AY727910.1 | - | 22 | 2687 | |||
AY727911.1 | - | 22 | 2590 | |||
AY727912.1 | - | 21 | 2860 | |||
AY727913.1 | - | 22 | 2687 | |||
BX648324.1 | - | 22 | 2657 | |||
CR933663.1 | - | 21 | 2501 |