Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
siRNA ID
HSS124072
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DZANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099407.1
NP_001092877.1
18
2070
XM_005260742.3
XP_005260799.1
18
2122
XM_005260743.3
XP_005260800.1
17
1976
XM_006723584.2
XP_006723647.1
16
1897
XM_011529268.2
XP_011527570.1
18
2240
XM_011529271.2
XP_011527573.1
18
2853
XM_011529272.2
XP_011527574.1
18
2236
XM_011529275.2
XP_011527577.1
17
2726
XM_011529277.2
XP_011527579.1
10
2967
XM_017027909.1
XP_016883398.1
18
2223
XM_017027910.1
XP_016883399.1
18
2106
XM_017027911.1
XP_016883400.1
18
2102
XM_017027912.1
XP_016883401.1
18
2809
XM_017027913.1
XP_016883402.1
18
2794
XM_017027914.1
XP_016883403.1
18
2048
XM_017027915.1
XP_016883404.1
18
2044
XM_017027916.1
XP_016883405.1
16
2608
XM_017027917.1
XP_016883406.1
19
2903
XM_017027918.1
XP_016883407.1
19
2157
XM_017027919.1
XP_016883408.1
19
2153
XM_017027920.1
XP_016883409.1
17
2112
XM_017027921.1
XP_016883410.1
11
1274
GenBank mRNA
AK001462.1
-
12
1257
AK026306.1
-
4
279
AK055544.1
-
17
1865
AK131451.1
-
20
2893
AK295073.1
-
18
1980
AK299746.1
-
18
1993
BC130341.1
-
17
1898
40146372
BC144153.1
-
17
1898
9052673
CR749327.1
-
19
2122
DQ104738.1
-
9
991
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy in response to mitochondrial depolarization
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0