Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@393b71ef
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other ECHDC2 SIRNA Assays ›
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 52895910 - 52927212 on Build GRCh38
UniGene ID:
Hs.476319
Species:
Human
siRNA ID
HSS124258
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECHDC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198961.1
NP_001185890.1
2
249
NM_001198962.1
NP_001185891.1
2
249
NM_001319958.1
NP_001306887.1
3
762
NM_018281.3
NP_060751.2
2
249
XM_011541709.2
XP_011540011.1
3
460
XM_011541710.2
XP_011540012.1
3
460
XM_011541711.2
XP_011540013.1
3
460
XM_011541712.2
XP_011540014.1
3
460
XM_011541713.2
XP_011540015.1
2
4237
XM_011541715.2
XP_011540017.1
3
685
XM_011541719.2
XP_011540021.1
3
1682
XM_011541720.2
XP_011540022.1
2
4006
XM_011541721.2
XP_011540023.1
4
699
XM_011541722.2
XP_011540024.1
3
317
XM_011541723.2
XP_011540025.1
3
417
XM_011541724.2
XP_011540026.1
3
460
XM_011541725.2
XP_011540027.1
3
460
XM_011541726.2
XP_011540028.1
2
4237
XM_011541727.2
XP_011540029.1
2
4006
XM_017001638.1
XP_016857127.1
4
2071
XM_017001639.1
XP_016857128.1
3
476
XM_017001640.1
XP_016857129.1
2
639
XM_017001641.1
XP_016857130.1
4
1840
XM_017001642.1
XP_016857131.1
2
4237
GenBank mRNA
AK001810.1
-
2
201
AK299834.1
-
3
418
AK300896.1
-
3
288
AK302285.1
-
5
828
AK307381.1
-
2
144
AK310650.1
-
3
381
AK315906.1
-
3
380
BC038389.1
-
3
680
BC044574.1
-
2
179
5314083
BC051887.1
-
2
237
CF127686.1
-
2
193
Back To Top
More Information
Panther Classification:
Molecular Function -
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-blue:8080/10.253.228.88:8080.
git-commit: 578676318d66c763e4a01d7f975a961cd389eaca
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.18.0-2021.03.09-1.0